Learn More
Medchemexpress LLC HY-139664 5mg , GSK-3685032 CAS:2170137-61-6 Purity:>98%
SDP
Catalog No. 502258992
Encompass_Preferred
HY-139664 5mg GSK-3685032 CAS: 2170137-61-6 GSK-3685032 is a non-time-dependent, noncovalently, first-in-class reversible DNMT1-selective inhibitor, with an IC50 of 0.036 μM. GSK-3685032 induces robust loss of DNA methylation, transcriptional activation, and cancer cell growth inhibition. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-8992 Supplier Medchemexpress LLC Supplier No. HY1396645MG
HY-139664 5mg GSK-3685032 CAS: 2170137-61-6 GSK-3685032 is a non-time-dependent, noncovalently, first-in-class reversible DNMT1-selective inhibitor, with an IC50 of 0.036 μM. GSK-3685032 induces robust loss of DNA methylation, transcriptional activation, and cancer cell growth inhibition. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.