Learn More
Medchemexpress LLC GSK-3beta inhibitor 3 | 1448990-73-5 | MFCD34368538 | 99.2% | C18H14FNO2S | 10MG
SDP
Catalog No. 5000258694
Encompass_Preferred
GSK-3β inhibitor 3 is a covalent, irreversible small-molecule inhibitor of glycogen synthase kinase 3β used in biochemical and cellular research. It is described as potent and selective, with a reported IC50 of 6.6 μM, and is supplied as a high-purity solid suitable for mechanistic and disease-model studies.
  • Covalent, irreversible inhibitor of GSK-3β for mechanistic studies.
  • Reported IC50 of 6.6 μM for activity benchmarking.
  • High purity (99.2%) suitable for biochemical assays and compound screening.
  • Solid, white to off-white form for straightforward handling and storage.
  • Available in small mg-scale quantities and as a DMSO solution for assay-ready use.
Catalog No. 50-002-58694 Supplier Medchemexpress LLC Supplier No. HY14148010MG
GSK-3β inhibitor 3 is a covalent, irreversible small-molecule inhibitor of glycogen synthase kinase 3β used in biochemical and cellular research. It is described as potent and selective, with a reported IC50 of 6.6 μM, and is supplied as a high-purity solid suitable for mechanistic and disease-model studies.
  • Covalent, irreversible inhibitor of GSK-3β for mechanistic studies.
  • Reported IC50 of 6.6 μM for activity benchmarking.
  • High purity (99.2%) suitable for biochemical assays and compound screening.
  • Solid, white to off-white form for straightforward handling and storage.
  • Available in small mg-scale quantities and as a DMSO solution for assay-ready use.