Learn More
Enzo Life Sciences GSK-3beta Inhibitor XVIII (5mg). CAS: 1139875-74-3
SDP
Catalog No. 502013428
Encompass_Preferred
Alternative name: 2-(Chloro-4-(4-thiophen-2-yl-pyrimidin-2-ylamino)-phenyl)-(4-methyl-pierazin-1-yl)-methanone. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3428 Supplier Enzo Life Sciences Supplier No. ENZCHM1340005
Alternative name: 2-(Chloro-4-(4-thiophen-2-yl-pyrimidin-2-ylamino)-phenyl)-(4-methyl-pierazin-1-yl)-methanone. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.