Learn More
Enzo Life Sciences GSK-3beta polyclonal antibody (25µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPST002C
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of rat GSK-3β. The sequence is completely conserved in human. UniProt ID: P18266. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Bovine. Applications: ICC, IHC (PS), WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7295