Learn More
MilliporeSigma GSK-3BETA RECOM ENZYME
Catalog No. NC9690212
Encompass
GSK3 beta active Serine/Threonine Phosphorylation 5mcg 125mcg 250mcg Protein Phosphorylation Assay: 5-40ng PURITY:>95% by SDS-PAGE STABILITY: 6 months at - 20degreeC PROPERTIES: Specific activity guaranteed between 932 and 1269U/mg
Catalog No. NC9690212 Supplier MilliporeSigma Supplier No. 14306
GSK3 beta active Serine/Threonine Phosphorylation 5mcg 125mcg 250mcg Protein Phosphorylation Assay: 5-40ng PURITY:>95% by SDS-PAGE STABILITY: 6 months at - 20degreeC PROPERTIES: Specific activity guaranteed between 932 and 1269U/mg