Learn More
Medchemexpress LLC HY-101027 100mg Medchemexpress, GSK 4027 CAS:2079896-25-4 Purity:>98%
SDP
Catalog No. 501932013
Encompass_Preferred
Medchemexpress, HY-101027 100mg GSK 4027 CAS:2079896-25-4 GSK 4027 is a chemical probe for the PCAF / GCN5 bromodomain with an pIC 50 of 7.4±0.11 for PCAF in a time-resolved fluorescence resonance energy transfer (TR-FRET) assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2013 Supplier Medchemexpress LLC Supplier No. HY101027100MG
Add to Cart
Add to Cart
Medchemexpress, HY-101027 100mg GSK 4027 CAS:2079896-25-4 GSK 4027 is a chemical probe for the PCAF / GCN5 bromodomain with an pIC 50 of 7.4±0.11 for PCAF in a time-resolved fluorescence resonance energy transfer (TR-FRET) assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.