Learn More
Apexbio Technology LLC GSK 4716 101574-65-6 10mg
SDP
Catalog No. 502240812
Encompass_Preferred
GSK 4716 (CAS 101574-65-6) is a small-molecule agonist targeting estrogen-related receptors ERR and ERR It is designed to activate these nuclear receptors thereby modulating mitochondrial function and cellular energy metabolic processes GSK 4716 exerts its biological activity primarily through nuclear receptor-mediated gene regulation pathways Based on these pharmacological properties GSK 4716 holds research potential in investigating mitochondrial bioenergetics metabolic regulation and pathophysiological conditions related to metabolic disorders
This item is not returnable. View return policy
Catalog No. 50-224-0812 Supplier Apexbio Technology LLC Supplier No. B726510
GSK 4716 (CAS 101574-65-6) is a small-molecule agonist targeting estrogen-related receptors ERR and ERR It is designed to activate these nuclear receptors thereby modulating mitochondrial function and cellular energy metabolic processes GSK 4716 exerts its biological activity primarily through nuclear receptor-mediated gene regulation pathways Based on these pharmacological properties GSK 4716 holds research potential in investigating mitochondrial bioenergetics metabolic regulation and pathophysiological conditions related to metabolic disorders