Learn More
Medchemexpress LLC HY-13032A 5mg Medchemexpress, GSK 525768A CAS:1260530-25-3 Purity:>98%
SDP
Catalog No. 502081235
Encompass_Preferred
Medchemexpress, HY-13032A 5mg GSK 525768A CAS:1260530-25-3 GSK 525768A is the inactive enantiomer of GSK525762A. GSK 525768A has no activity towards BET. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1235 Supplier Medchemexpress LLC Supplier No. HY13032A5MG
Medchemexpress, HY-13032A 5mg GSK 525768A CAS:1260530-25-3 GSK 525768A is the inactive enantiomer of GSK525762A. GSK 525768A has no activity towards BET. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.