Learn More
Selleck Chemical LLC GSK'547 2mg 2226735-55-1
SDP
Supplier:  Selleck Chemical LLC S87872MG
Encompass_Preferred
GSK'547 is a highly selective and potent inhibitor of RIP1 (RIPK1) exhibiting a 400-fold improvement in mouse pharmacokinetic oral exposure compared with GSK'963 *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8385