Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSK-5959 | 901245-65-6 | 99.7% | 394.47 | 5 MG
SDP

Catalog No. 5000393332
Encompass_Preferred

GSK-5959 is a potent, selective, and cell-permeable BRPF1 bromodomain inhibitor. It exhibits an IC50 of approximately 80 nM, making it suitable for laboratory research.

  • Potent BRPF1 bromodomain inhibitor
  • Selective and cell-permeable
  • IC50 of approximately 80 nM
  • Purity of 99.73% (LCMS)
  • Appears as a light yellow to yellow solid
  • Molecular formula: C22H26N4O3
  • Molecular weight: 394.47

Catalog No. 50-003-93332 Supplier Medchemexpress LLC Supplier No. 5MGHY18665
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK-5959 is a potent, selective, and cell-permeable BRPF1 bromodomain inhibitor. It exhibits an IC50 of approximately 80 nM, making it suitable for laboratory research.

  • Potent BRPF1 bromodomain inhibitor
  • Selective and cell-permeable
  • IC50 of approximately 80 nM
  • Purity of 99.73% (LCMS)
  • Appears as a light yellow to yellow solid
  • Molecular formula: C22H26N4O3
  • Molecular weight: 394.47

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.