Learn More
Medchemexpress LLC HY-105309 10mg Medchemexpress, GSK-626616 CAS:1025821-33-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10530910MG
Encompass_Preferred
Medchemexpress, HY-105309 10mg GSK-626616 CAS:1025821-33-3 GSK-626616 is a potent, orally bioavailable inhibitor of DYRK3 (IC50=0.7 nM). GSK-626616 inhibits other members of the DYRK family (e.g., DYRK1A and DYRK2) with similar potency, which is a potential therapy for the treatment of anemia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8267