Learn More
Medchemexpress LLC HY-105309 5mg Medchemexpress, GSK-626616 CAS:1025821-33-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1053095MG
Encompass_Preferred
Medchemexpress, HY-105309 5mg GSK-626616 CAS:1025821-33-3 GSK-626616 is a potent, orally bioavailable inhibitor of DYRK3 ( IC 50 =0.7 nM). GSK-626616 inhibits other members of the DYRK family (e.g., DYRK1A and DYRK2) with similar potency, which is a potential therapy for the treatment of anemia [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2262