Learn More
Apexbio Technology LLC GSK 650394 890842-28-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062092
Encompass_Preferred
GSK 650394 is a small-molecule inhibitor selectively targeting serum- and glucocorticoid-regulated kinase 1 (SGK1) showing inhibition through competitive binding mechanism with an IC50 of approximately 13 nM in fluorescence polarization assays SGK1 functions as an androgen-regulated kinase inhibition of this signaling reduces androgen-driven growth in prostate cancer cell lines Experimental studies demonstrate that GSK 650394 blocks SGK1 and SGK2 kinase activity with IC50 values of 62 nM and 103 nM respectively assessed by scintillation proximity assays Additionally GSK 650394 reduces androgen-induced phosphorylation of SGK substrate Nedd4-2 and suppresses androgen-stimulated proliferation of prostate cancer LNCaP cells (IC50 1 M) suggesting its use in prostate cancer research as a potential therapeutic strategy
Catalog No. 50-000-62092 Supplier Apexbio Technology LLC Supplier No. B10515.1
GSK 650394 is a small-molecule inhibitor selectively targeting serum- and glucocorticoid-regulated kinase 1 (SGK1) showing inhibition through competitive binding mechanism with an IC50 of approximately 13 nM in fluorescence polarization assays SGK1 functions as an androgen-regulated kinase inhibition of this signaling reduces androgen-driven growth in prostate cancer cell lines Experimental studies demonstrate that GSK 650394 blocks SGK1 and SGK2 kinase activity with IC50 values of 62 nM and 103 nM respectively assessed by scintillation proximity assays Additionally GSK 650394 reduces androgen-induced phosphorylation of SGK substrate Nedd4-2 and suppresses androgen-stimulated proliferation of prostate cancer LNCaP cells (IC50 1 M) suggesting its use in prostate cancer research as a potential therapeutic strategy