Learn More
Medchemexpress LLC HY-117226A 10mg Medchemexpress, GSK 690 (Hydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY117226A10MG
Encompass_Preferred
Medchemexpress, HY-117226A 10mg GSK 690 (HCl) CAS: GSK 690 (Hydrochloride) is a reversible inhibitor of lysine specific demethylase 1 (LSD1), with a Kd value of 9 nM and a biochemical IC50 of 37 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9458