Learn More
Medchemexpress LLC GSK 690 Hydrochlorid 100mg | 2436760-79-9 | 405.92 | C24H24ClN3O | 100 MG
SDP
Catalog No. 5000226777
Encompass_Preferred
GSK 690 hydrochloride is a reversible small-molecule inhibitor of lysine-specific demethylase 1 (LSD1) with a reported Kd of 9 nM and a biochemical IC50 of 37 nM. It is supplied as a light yellow to yellow solid (CAS 2436760-79-9) with reported purity of 98.58% and high solubility in DMSO.
Catalog No. 50-002-26777 Supplier Medchemexpress LLC Supplier No. HY117226A100MG
GSK 690 hydrochloride is a reversible small-molecule inhibitor of lysine-specific demethylase 1 (LSD1) with a reported Kd of 9 nM and a biochemical IC50 of 37 nM. It is supplied as a light yellow to yellow solid (CAS 2436760-79-9) with reported purity of 98.58% and high solubility in DMSO.