Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSK 690 Hydrochlorid 10mM 1mL | 2436760-79-9 | 405.92 | C24H24ClN3O | 1 ML
SDP

Catalog No. 5000228509
Encompass_Preferred

GSK 690 Hydrochloride is a reversible inhibitor of lysine-specific demethylase 1 (LSD1) with reported Kd 9 nM and biochemical IC50 37 nM. It is supplied for research use in solution and solid formats and is characterized by high purity suitable for biochemical assays.

  • Reversible LSD1 inhibitor with Kd 9 nM and IC50 37 nM.
  • Supplied as 10 mM solution in DMSO (1 mL) and as solid quantities.
  • High purity (98.58%) suitable for biochemical and cell-based studies.
  • Molecular weight 405.92 and formula C24H24ClN3O.
  • Storage: sealed at 4°C; in solvent: -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-28509 Supplier Medchemexpress LLC Supplier No. HY117226A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK 690 Hydrochloride is a reversible inhibitor of lysine-specific demethylase 1 (LSD1) with reported Kd 9 nM and biochemical IC50 37 nM. It is supplied for research use in solution and solid formats and is characterized by high purity suitable for biochemical assays.

  • Reversible LSD1 inhibitor with Kd 9 nM and IC50 37 nM.
  • Supplied as 10 mM solution in DMSO (1 mL) and as solid quantities.
  • High purity (98.58%) suitable for biochemical and cell-based studies.
  • Molecular weight 405.92 and formula C24H24ClN3O.
  • Storage: sealed at 4°C; in solvent: -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.