Learn More
Abcam GSK-690693 hydrochloride, ATP-competitive pan-Akt kinase inhibitor, 25MG
Catalog No. 502672796
Encompass_Preferred
MW 425.5 Da, Purity >99%. Novel ATP-competitive pan-Akt kinase inhibitor (IC₅₀ values are 2, 13 and 9 nM for Akt1, 2 and 3 respectively). Also exhibits some inhibition of AMPK, PKA and PAK and PKC isoforms (IC₅₀ < 100 nM). Displays antiproliferative and apoptotic effects in tumor cell lines.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2796 Supplier Abcam Supplier No. AB21986925MG
MW 425.5 Da, Purity >99%. Novel ATP-competitive pan-Akt kinase inhibitor (IC₅₀ values are 2, 13 and 9 nM for Akt1, 2 and 3 respectively). Also exhibits some inhibition of AMPK, PKA and PAK and PKC isoforms (IC₅₀ < 100 nM). Displays antiproliferative and apoptotic effects in tumor cell lines.
The product is subject to the following: Abcam Restricted Use Statement