Learn More
Medchemexpress LLC HY-12507 5mg Medchemexpress, GSK-7975A CAS:1253186-56-9 Purity:>98%
SDP
Catalog No. 502081067
Encompass_Preferred
Medchemexpress, HY-12507 5mg GSK-7975A CAS:1253186-56-9 GSK-7975A is a potent and orally available CRAC channel inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1067 Supplier Medchemexpress LLC Supplier No. HY125075MG
Medchemexpress, HY-12507 5mg GSK-7975A CAS:1253186-56-9 GSK-7975A is a potent and orally available CRAC channel inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.