Learn More
Medchemexpress LLC HY-101872 10mg Medchemexpress, GSK-872 CAS:1346546-69-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10187210MG
Encompass_Preferred
Medchemexpress, HY-101872 10mg GSK-872 CAS:1346546-69-7 GSK-872 is a RIPK3 inhibitor, which binds RIP3 kinase domain with an IC50 of 1.8 nM, and inhibits kinase activity with an IC50 of 1.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8686