Learn More
Medchemexpress LLC GSK-872 | 1346546-69-7 | MFCD30481302 | 99.8% | 383.49 | C19H17N3O2S2 | 100 MG
SDP
Catalog No. 5000248037
Encompass_Preferred
GSK-872 is a potent, selective small-molecule inhibitor of receptor-interacting serine/threonine-protein kinase 3 (RIPK3) supplied for research use. It is intended for biochemical and cellular studies of necroptosis and RIPK3-dependent signaling and is provided as a characterized, high-purity solid.
  • Potent RIPK3 inhibition (IC50 ≈ 1.8 nM).
  • Suitable for biochemical and cell-based assays.
  • High purity for reproducible results.
  • Soluble in DMSO for convenient formulation.
  • Well characterized molecular properties for identification.
Catalog No. 50-002-48037 Supplier Medchemexpress LLC Supplier No. HY101872100MG
GSK-872 is a potent, selective small-molecule inhibitor of receptor-interacting serine/threonine-protein kinase 3 (RIPK3) supplied for research use. It is intended for biochemical and cellular studies of necroptosis and RIPK3-dependent signaling and is provided as a characterized, high-purity solid.
  • Potent RIPK3 inhibition (IC50 ≈ 1.8 nM).
  • Suitable for biochemical and cell-based assays.
  • High purity for reproducible results.
  • Soluble in DMSO for convenient formulation.
  • Well characterized molecular properties for identification.