Learn More
Medchemexpress LLC Gsk-872 | 1346546-69-7 | MFCD30481302 | 99.9% | 383.49 g·mol⁻¹ | C19H17N3O2S2 | 1 ML
SDP
Catalog No. 5000248178
Encompass_Preferred
GSK-872 is a potent and selective inhibitor of receptor-interacting protein kinase 3 (RIPK3), provided for research use as a 10 mM solution in DMSO (1 mL). It is commonly used to investigate necroptosis and RIPK3-dependent signaling pathways in cellular studies.
  • Potent RIPK3 inhibition with low-nanomolar IC50.
  • High reported purity (~99.9%).
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • Stable storage recommendations provided for powder and solution formats.
  • Product documentation includes a data sheet and safety data sheet (SDS).
Catalog No. 50-002-48178 Supplier Medchemexpress LLC Supplier No. HY10187210MM 1ML
GSK-872 is a potent and selective inhibitor of receptor-interacting protein kinase 3 (RIPK3), provided for research use as a 10 mM solution in DMSO (1 mL). It is commonly used to investigate necroptosis and RIPK3-dependent signaling pathways in cellular studies.
  • Potent RIPK3 inhibition with low-nanomolar IC50.
  • High reported purity (~99.9%).
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • Stable storage recommendations provided for powder and solution formats.
  • Product documentation includes a data sheet and safety data sheet (SDS).