Learn More
Apexbio Technology LLC GSK-923295 1088965-37-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062813
Encompass_Preferred
GSK-923295 (CAS 1088965-37-0) is a highly potent inhibitor targeting the centromere-associated protein E (CENP-E) a kinesin motor protein essential for proper chromosome alignment and progression from metaphase to anaphase during cell division By blocking microtubule-stimulated ATPase activity of CENP-E GSK-923295 stabilizes an ATP-bound intermediate resulting in significantly reduced enzymatic function (IC50 3 2 nM) In cellular models GSK-923295 induces mitotic arrest with morphological changes similar to CENP-E knockdown In mouse xenograft models using Colo205 colon carcinoma cells administration of GSK-923295 leads to tumor growth inhibition and increased apoptosis supporting its utility in cancer research
Catalog No. 50-000-62813 Supplier Apexbio Technology LLC Supplier No. A34505.1
GSK-923295 (CAS 1088965-37-0) is a highly potent inhibitor targeting the centromere-associated protein E (CENP-E) a kinesin motor protein essential for proper chromosome alignment and progression from metaphase to anaphase during cell division By blocking microtubule-stimulated ATPase activity of CENP-E GSK-923295 stabilizes an ATP-bound intermediate resulting in significantly reduced enzymatic function (IC50 3 2 nM) In cellular models GSK-923295 induces mitotic arrest with morphological changes similar to CENP-E knockdown In mouse xenograft models using Colo205 colon carcinoma cells administration of GSK-923295 leads to tumor growth inhibition and increased apoptosis supporting its utility in cancer research