Learn More
Apexbio Technology LLC GSK-923295, 5mg. Cas: 1088965-37-0 MFCD: MFCD16038931. CENP-E inhibitor,small-molecule
SDP
Supplier:  Apexbio Technology LLC A34505
Encompass_Preferred
GSK923295 is a specific inhibitor of CENP-E kinesin motor with IC50 value of 3.2 nM [1].Centromere-associated protein E (CENP-E) is a mitotic kinesin, which is the connect of mitosis process with the mitotic checkpoint signaling. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4805