Learn More
Selleck Chemical LLC GSK'963 5mg 2049868-46-2
SDP
Supplier:  Selleck Chemical LLC S86425MG
Encompass_Preferred
GSK'963 is a chiral small-molecule inhibitor of RIP1 kinase (RIPK1) with an IC50 of 29 nM in FP binding assays. It is >10?000-fold selective for RIP1 over 339 other kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8158