Learn More
Cayman Chemical GSK-J1 (sodium salt), Purity: ≥95%, CAS: 1797832-71-3, 5 mg
SDP
Catalog No. 502954805
Encompass_Preferred
A potent, cell impermeable inhibitor of the H3K27 histone demethylases JMJD3 and UTX (IC50s = 18 and 56 µM, respectively as measured by mass spectrometry; IC50 = 60 nM in JMJD3 antibody-based assays)
Catalog No. 50-295-4805 Supplier Cayman Chemical Supplier No. 120545 MG
A potent, cell impermeable inhibitor of the H3K27 histone demethylases JMJD3 and UTX (IC50s = 18 and 56 µM, respectively as measured by mass spectrometry; IC50 = 60 nM in JMJD3 antibody-based assays)