Learn More
Medchemexpress LLC HY-15648A 10mg Medchemexpress, GSK-J2 CAS:1394854-52-4 Purity:>98%
SDP
Catalog No. 501969948
Encompass_Preferred
Medchemexpress, HY-15648A 10mg GSK-J2 CAS:1394854-52-4 GSK-J2 is an isomer of GSK-J1 that does not have any specific activity. GSK-J1 is a potent inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9948 Supplier Medchemexpress LLC Supplier No. HY15648A10MG
Medchemexpress, HY-15648A 10mg GSK-J2 CAS:1394854-52-4 GSK-J2 is an isomer of GSK-J1 that does not have any specific activity. GSK-J1 is a potent inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.