Learn More
Medchemexpress LLC HY-15648F 10mg Medchemexpress, GSK-J4 (hydrochloride) CAS:1797983-09-5 Purity:>98%
SDP
Catalog No. 501969949
Encompass_Preferred
Medchemexpress, HY-15648F 10mg GSK-J4 (HCl) CAS:1797983-09-5 GSK-J4 hydrochloride is a potent dual inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A with IC50s of 8.6 and 6.6 μM, respectively. GSK-J4 hydrochloride inhibits LPS-induced TNF-α production in human primary macrophages with an IC50 of 9 μM. GSK-J4 hydrochloride is a cell permeable prodrug of GSK-J1[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9949 Supplier Medchemexpress LLC Supplier No. HY15648F10MG
Medchemexpress, HY-15648F 10mg GSK-J4 (HCl) CAS:1797983-09-5 GSK-J4 hydrochloride is a potent dual inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A with IC50s of 8.6 and 6.6 μM, respectively. GSK-J4 hydrochloride inhibits LPS-induced TNF-α production in human primary macrophages with an IC50 of 9 μM. GSK-J4 hydrochloride is a cell permeable prodrug of GSK-J1[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.