Learn More
Selleck Chemical LLC GSK J4 HCl
SDP
Catalog No. 5000002028
Encompass_Preferred
GSK J4 HCl is a cell permeable prodrug of GSK J1 which is the first selective inhibitor of the H3K27 histone demethylase JMJD3 and UTX with IC50 of 60 nM in a cell-free assay and inactive against a panel of demethylases of the JMJ family
Catalog No. 50-000-02028 Supplier Selleck Chemical LLC Supplier No. S707050MG
GSK J4 HCl is a cell permeable prodrug of GSK J1 which is the first selective inhibitor of the H3K27 histone demethylase JMJD3 and UTX with IC50 of 60 nM in a cell-free assay and inactive against a panel of demethylases of the JMJ family