Learn More
Apexbio Technology LLC GSK J5 HCl, 5mg CAS# 1394854-51-3 (free base)
SDP
Catalog No. 501998406
Encompass_Preferred
GSK J5 HCl is the hydrochloride form of GSK J5. GSK J5 is an inactive isomer of GSK J4 and a cell-permeable ester derivative of inactive control GSK J2. Lysine-specific demethylase 6B (KDM6B), also known as Jumonji domain-containing protein D3 (JMJD3), was overexpressed in patients with AML and these patients have a poor prognosis. KDM6B-specific pharmacological inhibitor GSK-J4 had a significant anti-proliferative effect in AML cell lines and freshly isolated BM monocytes (MNCs) from AML patients, while H3K27me3 levels were also increasing. GSK-J4 also caused apoptosis and cell cycle arrest in vitro, and reduced tumor burden in vivo in AML xenograft mouse models. It is worth noting that injection of GSK-J4 attenuated disease progression in a human AML xenograft mouse model. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8406 Supplier Apexbio Technology LLC Supplier No. A87925
Add to Cart
Add to Cart
GSK J5 HCl is the hydrochloride form of GSK J5. GSK J5 is an inactive isomer of GSK J4 and a cell-permeable ester derivative of inactive control GSK J2. Lysine-specific demethylase 6B (KDM6B), also known as Jumonji domain-containing protein D3 (JMJD3), was overexpressed in patients with AML and these patients have a poor prognosis. KDM6B-specific pharmacological inhibitor GSK-J4 had a significant anti-proliferative effect in AML cell lines and freshly isolated BM monocytes (MNCs) from AML patients, while H3K27me3 levels were also increasing. GSK-J4 also caused apoptosis and cell cycle arrest in vitro, and reduced tumor burden in vivo in AML xenograft mouse models. It is worth noting that injection of GSK-J4 attenuated disease progression in a human AML xenograft mouse model. Other sizes are also available. Please inqury us for quote.