Learn More
Abcam GSK-J5, Inactive control for GSK-J4, 10MG
Catalog No. 502705806
Encompass_Preferred
MW 453.97 g/mol, Purity >97%. Cell-permeable, inactive control for histone demethylase JMJD3/UTX inhibitor GSK-J4 (ab144395). Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5806 Supplier Abcam Supplier No. AB14439710MG
MW 453.97 g/mol, Purity >97%. Cell-permeable, inactive control for histone demethylase JMJD3/UTX inhibitor GSK-J4 (ab144395). Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement