Learn More
Apexbio Technology LLC GSK-LSD1 2HCl 2102933-95-7 5mg
SDP
Catalog No. 5000088938
Encompass_Preferred
GSK-LSD1 2HCl (CAS 2102933-95-7) is a selective irreversible inhibitor of lysine-specific demethylase 1A (LSD1) a flavin-dependent histone demethylase that modulates mono- and dimethylated lysine residues and is implicated in processes such as oocyte growth and tissue-specific differentiation GSK-LSD1 2HCl inhibits LSD1 with an IC50 of 16 nM and exhibits over 1000-fold selectivity against related FAD-utilizing enzymes including LSD2 MAO-A and MAO-B In cancer cell lines it demonstrates EC50 values below 5 nM for both gene expression modulation and growth inhibition GSK-LSD1 2HCl serves as a valuable chemical probe in epigenetics and cancer research
Catalog No. 50-000-88938 Supplier Apexbio Technology LLC Supplier No. B49835
GSK-LSD1 2HCl (CAS 2102933-95-7) is a selective irreversible inhibitor of lysine-specific demethylase 1A (LSD1) a flavin-dependent histone demethylase that modulates mono- and dimethylated lysine residues and is implicated in processes such as oocyte growth and tissue-specific differentiation GSK-LSD1 2HCl inhibits LSD1 with an IC50 of 16 nM and exhibits over 1000-fold selectivity against related FAD-utilizing enzymes including LSD2 MAO-A and MAO-B In cancer cell lines it demonstrates EC50 values below 5 nM for both gene expression modulation and growth inhibition GSK-LSD1 2HCl serves as a valuable chemical probe in epigenetics and cancer research