Learn More
Medchemexpress LLC HY-100546A 5mg Medchemexpress, GSK-LSD1 Dihydrochloride CAS:2102933-95-7 Purity:>98%
SDP
Catalog No. 501931879
Encompass_Preferred
Medchemexpress, HY-100546A 5mg GSK-LSD1 Dihydrochloride CAS:2102933-95-7 GSK-LSD1 Dihydrochloride is a potent, selective and irreversible lysine specific demethylase 1 ( LSD1 ) inhibitor with an IC 50 of 16 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1879 Supplier Medchemexpress LLC Supplier No. HY100546A5MG
Medchemexpress, HY-100546A 5mg GSK-LSD1 Dihydrochloride CAS:2102933-95-7 GSK-LSD1 Dihydrochloride is a potent, selective and irreversible lysine specific demethylase 1 ( LSD1 ) inhibitor with an IC 50 of 16 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.