Learn More
TARGETMOL CHEMICALS INC GSK-LSD1 dihydrochloride 10MG
SDP
Catalog No. 502877607
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. GSK-LSD1 dihydrochloride (GSK-LSD1 2HCl) is a specificity irreversible LSD1 inhibitor (IC50 16 nM). The selectivity of GSK-LSD1 for LSD1 is >1000 fold over other closely related FAD utilizing enzymes (i.e. MAO-A, LSD2, MAO-B). Purity 99.22%
Catalog No. 50-287-7607 Supplier TARGETMOL CHEMICALS INC Supplier No. T231510MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. GSK-LSD1 dihydrochloride (GSK-LSD1 2HCl) is a specificity irreversible LSD1 inhibitor (IC50 16 nM). The selectivity of GSK-LSD1 for LSD1 is >1000 fold over other closely related FAD utilizing enzymes (i.e. MAO-A, LSD2, MAO-B). Purity 99.22%