Learn More
Medchemexpress LLC GSK-LSD1 dihydrochloride | 2102933-95-7 | 99.5% | 289.24 | C14H22Cl2N2 | 10 MG
SDP
Catalog No. 5000212761
Encompass_Preferred
GSK-LSD1 dihydrochloride is a potent, selective, and irreversible inhibitor of lysine-specific demethylase 1 (LSD1/KDM1A) with an IC50 of 16 nM. Provided as a solid for research use, the compound has been used in cellular studies to modulate gene expression, induce autophagy markers, and inhibit proliferation in various cancer cell lines. Handle and store according to safety data sheet recommendations.
Catalog No. 50-002-12761 Supplier Medchemexpress LLC Supplier No. HY100546A10MG
GSK-LSD1 dihydrochloride is a potent, selective, and irreversible inhibitor of lysine-specific demethylase 1 (LSD1/KDM1A) with an IC50 of 16 nM. Provided as a solid for research use, the compound has been used in cellular studies to modulate gene expression, induce autophagy markers, and inhibit proliferation in various cancer cell lines. Handle and store according to safety data sheet recommendations.