Learn More
Medchemexpress LLC GSK-LSD1 dihydrochloride | 2102933-95-7 | 99.5% | 289.24 g·mol⁻1 | C14H22Cl2N2 | 50 MG
SDP
Catalog No. 5000211927
Encompass_Preferred
This dihydrochloride salt is a potent, selective, and irreversible inhibitor of lysine-specific demethylase 1 (LSD1), used as a research probe for epigenetic regulation and metabolic studies. It exhibits an IC50 of 16 nM and is supplied with high reported purity.
  • Potent LSD1 inhibition with IC50 of 16 nM.
  • High purity (≈99.5%).
  • Supplied as a dihydrochloride salt in solid and DMSO solution formats.
  • Suitable for epigenetic and metabolic research applications.
  • Available in small research pack sizes including 50 MG.
Catalog No. 50-002-11927 Supplier Medchemexpress LLC Supplier No. HY100546A50MG
This dihydrochloride salt is a potent, selective, and irreversible inhibitor of lysine-specific demethylase 1 (LSD1), used as a research probe for epigenetic regulation and metabolic studies. It exhibits an IC50 of 16 nM and is supplied with high reported purity.
  • Potent LSD1 inhibition with IC50 of 16 nM.
  • High purity (≈99.5%).
  • Supplied as a dihydrochloride salt in solid and DMSO solution formats.
  • Suitable for epigenetic and metabolic research applications.
  • Available in small research pack sizes including 50 MG.