Learn More
Medchemexpress LLC GSK-LSD1 dihydrochloride | 2102933-95-7 | 99.5% | 289.24 g/mol | C14H22Cl2N2 | 1 ML
SDP
Catalog No. 5000213046
Encompass_Preferred
GSK-LSD1 dihydrochloride is a potent, selective, and irreversible inhibitor of lysine-specific demethylase 1 (LSD1) with an IC50 of 16 nM. It is intended for research use to probe LSD1 function, study gene expression changes, and evaluate effects on cancer cell proliferation. Supplied as a high-purity solid or as a solution in DMSO.
Catalog No. 50-002-13046 Supplier Medchemexpress LLC Supplier No. HY100546A10MM 1ML
GSK-LSD1 dihydrochloride is a potent, selective, and irreversible inhibitor of lysine-specific demethylase 1 (LSD1) with an IC50 of 16 nM. It is intended for research use to probe LSD1 function, study gene expression changes, and evaluate effects on cancer cell proliferation. Supplied as a high-purity solid or as a solution in DMSO.