Learn More
Cayman Chemical GSK-LSD1 (hydrochloride), 25 mg
SDP
Supplier:  Cayman Chemical 1643925
Encompass_Preferred
An irreversible, mechanism-based inhibitor of LSD1 (IC50 = 16 nM); induces gene expression changes in various cancer cell lines, inhibiting their proliferation (EC50s <5 nM)
Catalog No. NC0791305