Learn More
Selleck Chemical LLC GSK0660-5mg
SDP
Catalog No. 502390009
Encompass_Preferred
GSK0660 is a potent PPARβ/δ antagonist with a pIC50 of 6.8 (binding assay IC50 = 155 nM; antagonist assay IC50 = 300 nM) and is nearly inactive on PPARα and PPARγ with IC50s above approximately 10 μM.
Catalog No. 50-239-0009 Supplier Selleck Chemical LLC Supplier No. S58175MG
GSK0660 is a potent PPARβ/δ antagonist with a pIC50 of 6.8 (binding assay IC50 = 155 nM; antagonist assay IC50 = 300 nM) and is nearly inactive on PPARα and PPARγ with IC50s above approximately 10 μM.