Learn More
Medchemexpress LLC HY-12377 5mg Medchemexpress, GSK0660 CAS:1014691-61-2 Purity:>98%
SDP
Catalog No. 502081025
Encompass_Preferred
Medchemexpress, HY-12377 5mg GSK0660 CAS:1014691-61-2 GSK0660 is a potent antagonist of PPARβ and PPARδ, with IC50s of 155 nM for both isoforms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1025 Supplier Medchemexpress LLC Supplier No. HY123775MG
Medchemexpress, HY-12377 5mg GSK0660 CAS:1014691-61-2 GSK0660 is a potent antagonist of PPARβ and PPARδ, with IC50s of 155 nM for both isoforms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.