Learn More
Medchemexpress LLC HY-19608 10mg Medchemexpress, GSK1016790A CAS:942206-85-1 Purity:>98%
SDP
Catalog No. 502082148
Encompass_Preferred
Medchemexpress, HY-19608 10mg GSK1016790A CAS:942206-85-1 GSK1016790A is a potent and selective transient receptor potential vanilloid 4 (TRPV4) channel agonist. GSK1016790A can elicit Ca2+ influx and elevate intracellular Ca2+ in HEK cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2148 Supplier Medchemexpress LLC Supplier No. HY1960810MG
Medchemexpress, HY-19608 10mg GSK1016790A CAS:942206-85-1 GSK1016790A is a potent and selective transient receptor potential vanilloid 4 (TRPV4) channel agonist. GSK1016790A can elicit Ca2+ influx and elevate intracellular Ca2+ in HEK cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.