Learn More
Selleck Chemical LLC GSK1016790A 5mg 942206-85-1 GSK101
SDP
Supplier:  Selleck Chemical LLC S81075MG
Encompass_Preferred
GSK1016790A (GSK101) is a novel, potent activator of TRPV4 (transient receptor potential vanilloid 4) with EC50 of 34nM in choroid plexus epithelial cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5664