Learn More
Medchemexpress LLC HY-12036 5mg Medchemexpress, GSK1059615 CAS:958852-01-2 Purity:>98%
SDP
Catalog No. 502080937
Encompass_Preferred
Medchemexpress, HY-12036 5mg GSK1059615 CAS:958852-01-2 GSK1059615 is a dual inhibitor of PI3Kα/β/δ/γ (reversible) and mTOR with IC50 of 0.4 nM/0.6 nM/2 nM/5 nM and 12 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0937 Supplier Medchemexpress LLC Supplier No. HY120365MG
Medchemexpress, HY-12036 5mg GSK1059615 CAS:958852-01-2 GSK1059615 is a dual inhibitor of PI3Kα/β/δ/γ (reversible) and mTOR with IC50 of 0.4 nM/0.6 nM/2 nM/5 nM and 12 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.