Learn More
Selleck Chemical LLC GSK1059615 10mg 958852-01-2
SDP
Supplier:  Selleck Chemical LLC S136010MG
Encompass_Preferred
GSK1059615 is a dual inhibitor of PI3K?/?/?/? (reversible) and mTOR with IC50 of 0.4 nM/0.6 nM/2 nM/5 nM and 12 nM, respectively. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-749-9