Learn More
Cayman Chemical GSK106hydrochlorIde 10mg
SDP
Catalog No. 502965123
Encompass_Preferred
An inactive control for the selective PAD4 inhibitors, GSK484 and GSK199 (IC50s >100 µM)
Catalog No. 50-296-5123 Supplier Cayman Chemical Supplier No. 1749010 MG
An inactive control for the selective PAD4 inhibitors, GSK484 and GSK199 (IC50s >100 µM)