Learn More
Apexbio Technology LLC GSK1070916, 10mg. Cas: 942918-07-2 MFCD: MFCD16039223. Aurora B/C inhibitor
SDP
Supplier:  Apexbio Technology LLC A412710
Encompass_Preferred
GSK1070916 is a potent and selective inhibitor of Aurora B/C Kinase with Ki values of 0.38nM and 1.5nM, respectively [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4887