Learn More
Selleck Chemical LLC Trametinib (GSK1120212) 5mg 871700-17-3 JTP-74057, Mekinist
SDP
Supplier:  Selleck Chemical LLC S26735MG
Encompass_Preferred
Trametinib (GSK1120212, JTP-74057, Mekinist) is a highly specific and potent MEK1/2 inhibitor with IC50 of 0.92 nM/1.8 nM in cell-free assays, no inhibition of the kinase activities of c-Raf, B-Raf, ERK1/2. Trametinib activates autophagy and induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0370092