Learn More
Cayman Chemical GSK121 trifluoroacetate salt
SDP
Catalog No. 502968113
Encompass_Preferred
The initial compound identified in a screen for PAD4 inhibitors (IC50 = 3.2 µM in functional assay) that was optimized to produce the more potent PAD4 inhibitors, GSK484 and GSK199
Catalog No. 50-296-8113 Supplier Cayman Chemical Supplier No. 17491500 UG
The initial compound identified in a screen for PAD4 inhibitors (IC50 = 3.2 µM in functional assay) that was optimized to produce the more potent PAD4 inhibitors, GSK484 and GSK199