Learn More
Selleck Chemical LLC GSK126 5mg 1346574-57-9 GSK2816126A, GSK2816126
SDP
Supplier:  Selleck Chemical LLC S70615MG
Encompass_Preferred
GSK126 (GSK2816126A, GSK2816126) is a potent, highly selective EZH2 methyltransferase inhibitor with IC50 of 9.9 nM, >1000-fold selective for EZH2 over 20 other human methyltransferases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0944306