Learn More
Medchemexpress LLC HY-112398 10mg Medchemexpress, GSK1379725A CAS:1802251-00-8 Purity:>98%
SDP
Catalog No. 501932421
Encompass_Preferred
Medchemexpress, HY-112398 10mg GSK1379725A CAS:1802251-00-8 GSK1379725A is a selective BPTF ligand with a K d of 2.8 uM, showing no binding activity for Brd4 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2421 Supplier Medchemexpress LLC Supplier No. HY11239810MG
Medchemexpress, HY-112398 10mg GSK1379725A CAS:1802251-00-8 GSK1379725A is a selective BPTF ligand with a K d of 2.8 uM, showing no binding activity for Brd4 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.