Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSK143 dihydrochloride | 2341796-81-2 | MFCD28963898 | 99.4% | 415.32 g/mol | C17H24Cl2N6O2 | 5 MG
SDP

Catalog No. 5000264105
Encompass_Preferred

GSK143 dihydrochloride is the dihydrochloride salt of a research-active SYK inhibitor with demonstrated biochemical potency and cellular activity in hematologic models. It is supplied as a solid salt for laboratory research and biochemical assays.

  • Orally active, selective spleen tyrosine kinase (SYK) inhibitor.
  • Biochemical potency: SYK pIC50 = 7.5.
  • Cellular activity: IC50 in CLL cells = 323 nM.
  • Dihydrochloride salt; solid, off-white to light yellow appearance.
  • High purity (≈99.4%) suitable for research applications.
  • Molecular weight 415.32 g/mol; formula C17H24Cl2N6O2.

Catalog No. 50-002-64105 Supplier Medchemexpress LLC Supplier No. HY12736A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK143 dihydrochloride is the dihydrochloride salt of a research-active SYK inhibitor with demonstrated biochemical potency and cellular activity in hematologic models. It is supplied as a solid salt for laboratory research and biochemical assays.

  • Orally active, selective spleen tyrosine kinase (SYK) inhibitor.
  • Biochemical potency: SYK pIC50 = 7.5.
  • Cellular activity: IC50 in CLL cells = 323 nM.
  • Dihydrochloride salt; solid, off-white to light yellow appearance.
  • High purity (≈99.4%) suitable for research applications.
  • Molecular weight 415.32 g/mol; formula C17H24Cl2N6O2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.